Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate ≥99.0%

03842-1 03842-2 03842-3
103536-114EA 92.31 USD
103536-114 103536-116 103536-118
Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate ≥99.0%
Ethyl 6-chloroimidazo[1,2-b]pyridazine-3-carboxylate
Formula: C₉H₈ClN₃O₂
MDL Number: MFCD11975600
CAS Number: 1150566-27-0

Order Now

Specification Test Results

Purety 99.0%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR