5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine ≥98.4%

03806-1 03806-2 03806-3
103536-102EA 181.54 USD
103536-102 103536-104 103536-106
5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine ≥98.4%
5-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
MDL Number: MFCD11046240
CAS Number: 876343-82-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purety 98.4%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR