4-Iodo-1-methylimidazole ≥97.0%

Synonyms: 4-Iodo-1-methyl-1H-imidazole

03797-1 03797-2 03797-3
103535-074EA 38.46 USD
103535-074 103536-098 103536-100
4-Iodo-1-methylimidazole ≥97.0%
4-Iodo-1-methylimidazole
Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD01632214
CAS Number: 71759-87-0

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR