3-Bromo-2-chloro-4-methylpyridine ≥98.2%

03778-1 03778-2 03778-3
103535-068EA 30.77 USD
103535-068 103535-070 103535-072
3-Bromo-2-chloro-4-methylpyridine ≥98.2%
3-Bromo-2-chloro-4-methylpyridine
Formula: C₆H₅BrClN
MDL Number: MFCD04112511
CAS Number: 55404-31-4

Order Now

Specification Test Results

Purety 98.2%
Appearance Gray powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR