2-Amino-5-fluoropyridine ≥99.7%

03775-1 03775-2 03775-3
103535-062EA 30.77 USD
103535-062 103535-064 103535-066
2-Amino-5-fluoropyridine ≥99.7%
2-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD01861120
CAS Number: 21717-96-4

Order Now

Specification Test Results

Purety 99.7%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR