4-Amino-5-bromo-2-chloropyridine ≥96.0%

Synonyms: 5-bromo-2-chloropyridin-4-amine, 5-Bromo-2-chloro-4-pyridylamine, 5-Bromo-2-chloro-4-pyridinamine

03760-1 03760-2 03760-3
103535-056EA 50.77 USD
103535-056 103535-058 103535-060
4-Amino-5-bromo-2-chloropyridine ≥96.0%
4-Amino-5-bromo-2-chloropyridine
Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
MDL Number: MFCD09037459
CAS Number: 857730-21-3

Order Now

Specification Test Results

Purety 96.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR