6-Bromo-5-methoxy-1H-indazole ≥97.5%

03685-1 03685-2 03685-3
103535-050EA 115.38 USD
103535-050 103535-052 103535-054
6-Bromo-5-methoxy-1H-indazole ≥97.5%
6-Bromo-5-methoxy-1H-indazole
Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
MDL Number: MFCD16659026
CAS Number: 1206800-17-0

Order Now

Specification Test Results

Purety 97.5%
Appearance Brown powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR