6-Iodopyridazin-3-amine ≥98.0%

Synonyms: 3-Amino-6-iodopyridazine

03618-1 03618-2 03618-3
103535-044EA 61.54 USD
103535-044 103535-046 103535-048
6-Iodopyridazin-3-amine ≥98.0%
6-Iodopyridazin-3-amine
Formula: C₄H₄IN₃
MW: 221 g/mol
MDL Number: MFCD09991754
CAS Number: 187973-60-0

Order Now

Specification Test Results

Purety 98.0%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR