5-Chloro-1H-pyrazolo[4,3-b]pyridine ≥98.4%

03608-1 03608-2 03608-3
103535-038EA 56.92 USD
103535-038 103535-040 103535-042
5-Chloro-1H-pyrazolo[4,3-b]pyridine ≥98.4%
5-Chloro-1H-pyrazolo[4,3-b]pyridine
Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
MDL Number: MFCD05663984
CAS Number: 94220-45-8

Order Now

Specification Test Results

Purety 98.4%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR