Morpholin-3-one ≥97.0%

03555-1 03555-2 03555-3
103535-032EA 30.77 USD
103535-032 103535-034 103535-036
Morpholin-3-one ≥97.0%
Morpholin-3-one
Formula: C₄H₇NO₂
MDL Number: MFCD00631009
CAS Number: 109-11-5

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR