1H-Pyrazole-4-carboxylic acid ≥97.0%

Synonyms: 4-Pyrazolecarboxylic acid

03525-1 03525-2 03525-3
103535-026EA 100 USD
103535-026 103535-028 103535-030
1H-Pyrazole-4-carboxylic acid ≥97.0%
1H-Pyrazole-4-carboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
MDL Number: MFCD00011558
CAS Number: 37718-11-9

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR