(2-Chloro-3-pyridinyl)methanol ≥98.0%

Synonyms: 2-Chloro-3-pyridinemethanol, (2-chloropyridin-3-yl)methanol, 2-Chloro-3-pyridinylmethanol

03459-1 03459-2 03459-3
103535-014EA 30.77 USD
103535-014 103535-016 103535-018
(2-Chloro-3-pyridinyl)methanol ≥98.0%
(2-Chloro-3-pyridinyl)methanol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD01935961
CAS Number: 42330-59-6

Order Now

Specification Test Results

Purety 98.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR