Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥98.4%

Synonyms: Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate

03397-1 03397-2 03397-3
103534-996EA 18.46 USD
103534-996 103534-998 103535-000
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate ≥98.4%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Formula: C₈H₉ClN₂O₂S
MW: 232.69 g/mol
MDL Number: MFCD00006085
CAS Number: 5909-24-0

Order Now

Specification Test Results

Purety 98.4%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR