4-Bromophthalimide ≥97.9%

Synonyms: 5-Bromoisoindoline-1,3-dione, 5-bromo-2,3-dihydro-1H-isoindole-1,3-dione

03374-1 03374-2 03374-3
103534-972EA 30.77 USD
103534-972 103534-974 103534-976
4-Bromophthalimide ≥97.9%
4-Bromophthalimide
Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
MDL Number: MFCD00466049
CAS Number: 6941-75-9

Order Now

Specification Test Results

Purety 97.9%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR