2-Chloro-5-methoxypyrimidine ≥97.0%

03370-1 03370-2 03370-3
103534-966EA 23.08 USD
103534-966 103534-968 103534-970
2-Chloro-5-methoxypyrimidine ≥97.0%
2-Chloro-5-methoxypyrimidine
Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD08702770
CAS Number: 22536-65-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow solid,

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR