Thiophene-3-carbaldehyde ≥99.2%

Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde

03369-1 03369-2 03369-3
103534-960EA 30.77 USD
103534-960 103534-962 103534-964
Thiophene-3-carbaldehyde ≥99.2%
Thiophene-3-carbaldehyde
Formula: C₅H₄OS
MW: 112.15 g/mol
MDL Number: MFCD00005466
CAS Number: 498-62-4

Order Now

Specification Test Results

Purety 99.2%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR