1-Methyl-1H-pyrazole-5-carboxylic acid ≥98.6%

Synonyms: 1-Methyl-5-pyrazolecarboxylic acid

03362-1 03362-2 03362-3
103534-954EA 18.46 USD
103534-954 103534-956 103534-958
1-Methyl-1H-pyrazole-5-carboxylic acid ≥98.6%
1-Methyl-1H-pyrazole-5-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD00464253
CAS Number: 16034-46-1

Order Now

Specification Test Results

Purety 98.6%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR