1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥98.5%

Synonyms: 4-Oxopyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Ethyl ester, 1-tert-Butyl 3-ethyl 4-oxopyrrolidine-1,3-dicarboxylate

03321-1 03321-2 03321-3
103534-936EA 30.77 USD
103534-936 103534-938 103534-940
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥98.5%
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate
Formula: C₁₂H₁₉NO₅
MW: 257.2830 g/mol
MDL Number: MFCD06656201
CAS Number: 146256-98-6

Order Now

Specification Test Results

Purety 98.5%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR