6-Bromo-3-nitropyridin-2-amine ≥98.5%

Synonyms: 2-Amino-6-bromo-3-nitropyridine, 6-bromo-3-nitro-2-pyridinamine

03294-1 03294-2 03294-3
103534-924EA 49.23 USD
103534-924 103534-926 103534-928
6-Bromo-3-nitropyridin-2-amine ≥98.5%
6-Bromo-3-nitropyridin-2-amine
Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
MDL Number: MFCD07774108
CAS Number: 84487-04-7

Order Now

Specification Test Results

Purety 98.5%
Appearance Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR