cis-3-Amino-cyclobutanemethanol hydrochloride ≥97%

03187-01-1 03187-01-2 03187-01-3
103534-888EA 369.23 USD
103534-888 103534-890 103534-892
cis-3-Amino-cyclobutanemethanol hydrochloride ≥97%
cis-3-Amino-cyclobutanemethanol hydrochloride
Formula: C₅H₁₁NO·HCl
MW: 137.608 g/mol
CAS Number: 142733-65-1

Order Now

Specification Test Results

Purety 97%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR