3(2H)-Pyridazinone ≥97.0%

Synonyms: 2,3-dihydropyridazin-3-one, Pyridazin-3(2H)-one

03181-1 03181-2 03181-3
103534-882EA 12.31 USD
103534-882 103534-884 103534-886
3(2H)-Pyridazinone ≥97.0%
3(2H)-Pyridazinone
Formula: C₄H₄N₂O
MW: 96.0874 g/mol
MDL Number: MFCD00099605
CAS Number: 504-30-3

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR