3-Bromoisonicotinic acid ≥99.6%

Synonyms: 3-Bromopyridine-4-carboxylic acid, 3-Bromo-4-pyridinecarboxylic acid

03179-1 03179-2 03179-3
103534-876EA 30.77 USD
103534-876 103534-878 103534-880
3-Bromoisonicotinic acid ≥99.6%
3-Bromoisonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
MDL Number: MFCD00040944
CAS Number: 13959-02-9

Order Now

Specification Test Results

Purety 99.6%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR