5-Bromo-2,2-difluoro-1,3-benzodioxole ≥97.7%

Synonyms: 5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole

03161-1 03161-2 03161-3
103534-870EA 30.77 USD
103534-870 103534-872 103534-874
5-Bromo-2,2-difluoro-1,3-benzodioxole ≥97.7%
5-Bromo-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₃BrF₂O₂
MW: 237 g/mol
MDL Number: MFCD00236212
CAS Number: 33070-32-5

Order Now

Specification Test Results

Purety 97.7%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR