trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate ≥99.3%

Synonyms: tert-Butyl trans-3-fluoro-4-hydroxypyrrolidine-1-carboxylate, trans-3-Fluoro-4-hydroxypyrrolidine, N-BOC protected

03140-1 03140-2 03140-3
103534-864EA 92.31 USD
103534-864 103534-866 103534-868
trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate ≥99.3%
trans-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate
Formula: C₉H₁₆FNO₃
MW: 205.23 g/mol
MDL Number: MFCD18791200
CAS Number: 869481-93-6

Order Now

Specification Test Results

Purety 99.3%
Appearance Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR