tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate ≥97.8%

Synonyms: 1-Boc-4-Fluoro-4-(hydroxymethyl)piperidine

03119-1 03119-2 03119-3
103536-080EA 138.46 USD
103536-080 103536-082 103536-084
tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate ≥97.8%
tert-Butyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate
Formula: C₁₁H₂₀FNO₃
MW: 233.28 g/mol
MDL Number: MFCD08062513
CAS Number: 614730-97-1

Order Now

Specification Test Results

Purety 97.8%
Appearance Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR