3-Fluoropyridine-2-carbonitrile ≥97.0%

Synonyms: 3-Fluoro-2-pyridinecarbonitrile, 2-Cyano-3-fluoropyridine, 3-Fluoropicolinonitrile

03116-1 03116-2 03116-3
103534-860EA 30.77 USD
103534-860 103534-862 103536-078
3-Fluoropyridine-2-carbonitrile ≥97.0%
3-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD06797501
CAS Number: 97509-75-6

Order Now

Specification Test Results

Purety 97.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR