7-Nitro-1H-indazole ≥97.0%

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

03039-1 03039-2 03039-3
103534-842EA 46.15 USD
103534-842 103534-844 103534-846
7-Nitro-1H-indazole ≥97.0%
7-Nitro-1H-indazole
Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
MDL Number: MFCD00022789
CAS Number: 2942-42-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR