4-Methyloxan-4-amine hydrochloride ≥97%

Synonyms: 4-Methyltetrahydro-2H-pyran-4-amine hydrochloride

02970-1 02970-2 02970-3
103534-824EA 69.23 USD
103534-824 103534-826 103534-828
4-Methyloxan-4-amine hydrochloride ≥97%
4-Methyloxan-4-amine hydrochloride
Formula: C₆H₁₃NO·HCl
MW: 151.634 g/mol
MDL Number: MFCD13184221
CAS Number: 851389-38-3

Order Now

Specification Test Results

Purety 97%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR