5-Bromo-1H-pyrazolo[3,4-c]pyridine ≥99.6%

02945-1 02945-2 02945-3
103534-818EA 69.23 USD
103534-818 103534-820 103534-822
5-Bromo-1H-pyrazolo[3,4-c]pyridine ≥99.6%
5-Bromo-1H-pyrazolo[3,4-c]pyridine
Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD11518975
CAS Number: 929617-35-6

Order Now

Specification Test Results

Purety 99.6%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR