tert-Butyl (4-methylpiperidin-4-yl)carbamate ≥98.1%

Synonyms: tert-butyl N-(4-methylpiperidin-4-yl)carbamate

02909-1 02909-2 02909-3
103534-812EA 76.92 USD
103534-812 103534-814 103534-816
tert-Butyl (4-methylpiperidin-4-yl)carbamate ≥98.1%
tert-Butyl (4-methylpiperidin-4-yl)carbamate
Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
MDL Number: MFCD08703167
CAS Number: 163271-08-7

Order Now

Specification Test Results

Purety 98.1%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR