tert-Butyl n-(3-methylpiperidin-3-yl)carbamate ≥97%

Synonyms: tert-Butyl (3-methylpiperidin-3-yl)carbamate, 3-Amino-3-methylpiperidine, 3-BOC protected

02907-1 02907-2 02907-3
103534-806EA 153.85 USD
103534-806 103534-808 103534-810
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate ≥97%
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate
Formula: C₁₁H₂₂N₂O₂
MW: 214.305 g/mol
MDL Number: MFCD13194151
CAS Number: 169750-96-3

Order Now

Specification Test Results

Purety 97%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR