1-(Benzenesulfonyl)-4-chloro-1H-pyrrolo[2,3-b]pyridine ≥99.2%

Synonyms: 4-Chloro-1-benzenesulphonyl-7-azaindole, 4-Chloro-1-(phenylsulphonyl)-1H-pyrrolo[2,3-b]pyridine

02736-1 02736-2 02736-3
103534-800EA 30.77 USD
103534-800 103534-802 103534-804
1-(Benzenesulfonyl)-4-chloro-1H-pyrrolo[2,3-b]pyridine ≥99.2%
1-(Benzenesulfonyl)-4-chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₁₃H₉ClN₂O₂S
MW: 292.741 g/mol
MDL Number: MFCD04153750
CAS Number: 744209-63-0

Order Now

Specification Test Results

Purety 99.2%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR