1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid ≥98.5%

Synonyms: 1-(tert-Butoxycarbonyl)-3-methylazetidine-3-carboxylic acid

02648-1 02648-2 02648-3
103534-794EA 104.62 USD
103534-794 103534-796 103534-798
1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid ≥98.5%
1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid
Formula: C₁₀H₁₇NO₄
MW: 215.246 g/mol
MDL Number: MFCD12406581
CAS Number: 887591-62-0

Order Now

Specification Test Results

Purety 98.5%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR