1-tert-Butyl 3-methyl 3-methylazetidine-1,3-dicarboxylate ≥98.4%

02647-1 02647-2 02647-3
103534-788EA 123.08 USD
103534-788 103534-790 103534-792
1-tert-Butyl 3-methyl 3-methylazetidine-1,3-dicarboxylate ≥98.4%
1-tert-Butyl 3-methyl 3-methylazetidine-1,3-dicarboxylate
Formula: C₁₁H₁₉NO₄
CAS Number: 898228-37-0

Order Now

Specification Test Results

Purety 98.4%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR