1-Boc-4-methylpiperidine-4-carboxylic acid ≥97.0%

Synonyms: 1-Boc-4-Methylpiperidine-4-carboxylic acid, 1-[(tert-butoxy)carbonyl]-4-methylpiperidine-4-carboxylic acid

02572-1 02572-2 02572-3
103534-776EA 38.46 USD
103534-776 103534-778 103534-780
1-Boc-4-methylpiperidine-4-carboxylic acid ≥97.0%
1-Boc-4-methylpiperidine-4-carboxylic acid
Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD02179132
CAS Number: 189321-63-9

Order Now

Specification Test Results

Purety 97.0%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR