4-Amino-7H-pyrrolo[2,3-d]pyrimidine ≥97.0%

02551-1 02551-2 02551-3
103534-764EA 30.77 USD
103534-764 103534-766 103534-768
4-Amino-7H-pyrrolo[2,3-d]pyrimidine ≥97.0%
4-Amino-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₆H₆N₄
MDL Number: MFCD01686848
CAS Number: 1500-85-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR