2-Hydroxyisonicotinic acid ≥97.0%

Synonyms: 2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid

02487-1 02487-2 02487-3
103534-752EA 30.77 USD
103534-752 103534-754 103534-756
2-Hydroxyisonicotinic acid ≥97.0%
2-Hydroxyisonicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD01318566
CAS Number: 22282-72-0

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR