4-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one ≥97.5%

Synonyms: 4-chloro-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one

02409-1 02409-2 02409-3
103534-734EA 138.46 USD
103534-734 103534-736 103534-738
4-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one ≥97.5%
4-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formula: C₇H₅ClN₂O
MW: 168.5804 g/mol
MDL Number: MFCD11518939
CAS Number: 346599-62-0

Order Now

Specification Test Results

Purety 97.5%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR