1H-Indazole-3-carboxylic acid ≥97.0%

Synonyms: Indazole-3-carboxylic acid

02406-1 02406-2 02406-3
103534-728EA 30.77 USD
103534-728 103534-730 103534-732
1H-Indazole-3-carboxylic acid ≥97.0%
1H-Indazole-3-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00211066
CAS Number: 4498-67-3

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR