2-Aminonicotinic acid ≥97.9%

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

02391-1 02391-2 02391-3
103534-722EA 12.31 USD
103534-722 103534-724 103534-726
2-Aminonicotinic acid ≥97.9%
2-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
MDL Number: MFCD00006318
CAS Number: 5345-47-1

Order Now

Specification Test Results

Purety 97.9%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR