N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester ≥98.8%

Synonyms: N-Boc-trans-4-hydroxy-L-proline Methyl Ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-Hyp-OMe, 1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate

02363-1 02363-2 02363-3
103534-716EA 30.77 USD
103534-716 103534-718 103534-720
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester ≥98.8%
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
MDL Number: MFCD00076981
CAS Number: 74844-91-0

Order Now

Specification Test Results

Purety 98.8%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR