(S)-(+)-Prolinol ≥97.0%

Synonyms: (S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol

02356-1 02356-2 02356-3
103534-710EA 36.92 USD
103534-710 103534-712 103534-714
(S)-(+)-Prolinol ≥97.0%
(S)-(+)-Prolinol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD00005255
CAS Number: 23356-96-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Orange liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR