4-Bromo-1-methyl-1H-pyrazole ≥97.0%

02353-1 02353-2 02353-3
103534-704EA 30.77 USD
103534-704 103534-706 103534-708
4-Bromo-1-methyl-1H-pyrazole ≥97.0%
4-Bromo-1-methyl-1H-pyrazole
Formula: C₄H₅BrN₂
MW: 161.00 g/mol
MDL Number: MFCD02179565
CAS Number: 15803-02-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR