4-Amino-3-nitropyridine ≥98.6%

Synonyms: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

02338-1 02338-2 02338-3
103534-698EA 12.31 USD
103534-698 103534-700 103534-702
4-Amino-3-nitropyridine ≥98.6%
4-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD00160304
CAS Number: 1681-37-4

Order Now

Specification Test Results

Purety 98.6%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR