2-Methylthio-4-pyrimidinol ≥97.0%

Synonyms: 2-(Methylthio)pyrimidin-4(3H)-one, 2-(methylsulphanyl)-3,4-dihydropyrimidin-4-one, 2-Methylthio-4-pyrimidone, 4-Hydroxy-2-methylthiopyrimidine, S-Methyl-2-thiouracil

02320-1 02320-2 02320-3
103534-692EA 12.31 USD
103534-692 103534-694 103534-696
2-Methylthio-4-pyrimidinol ≥97.0%
2-Methylthio-4-pyrimidinol
Formula: C₅H₆N₂OS
MW: 142.18 g/mol
MDL Number: MFCD00047373
CAS Number: 5751-20-2

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR