Methyl (4-bromophenyl)acetate ≥98.6%

Synonyms: methyl 2-(4-bromophenyl)acetate

02296-1 02296-2 02296-3
103534-686EA 38.46 USD
103534-686 103534-688 103534-690
Methyl (4-bromophenyl)acetate ≥98.6%
Methyl (4-bromophenyl)acetate
Formula: C₉H₉BrO₂
MW: 229.0700 g/mol
MDL Number: MFCD00126844
CAS Number: 41841-16-1

Order Now

Specification Test Results

Purety 98.6%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR