5-Bromo-4-chloro-2-(methylthio)pyrimidine ≥98.5%

Synonyms: 5-bromo-4-chloro-2-(methylsulphanyl)pyrimidine

02264-1 02264-2 02264-3
103534-680EA 38.46 USD
103534-680 103534-682 103534-684
5-Bromo-4-chloro-2-(methylthio)pyrimidine ≥98.5%
5-Bromo-4-chloro-2-(methylthio)pyrimidine
Formula: C₅H₄BrClN₂S
MW: 239.5200 g/mol
MDL Number: MFCD00023232
CAS Number: 63810-78-6

Order Now

Specification Test Results

Purety 98.5%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR