4-Chloro-2-(methylthio)pyrimidine ≥97.0%

Synonyms: 4-Chloro-2-methylthiopyrimidine

02263-1 02263-2 02263-3
103534-674EA 30.77 USD
103534-674 103534-676 103534-678
4-Chloro-2-(methylthio)pyrimidine ≥97.0%
4-Chloro-2-(methylthio)pyrimidine
Formula: C₅H₅ClN₂S
MW: 160.63 g/mol
MDL Number: MFCD00006083
CAS Number: 49844-90-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR