Methyl-2-aminoisonicotinate ≥99.7%

Synonyms: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate

02229-1 02229-2 02229-3
103534-668EA 23.08 USD
103534-668 103534-670 103534-672
Methyl-2-aminoisonicotinate ≥99.7%
Methyl 2-aminoisonicotinate
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD04039316
CAS Number: 6937-03-7

Order Now

Specification Test Results

Purety 99.7%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR