4-Acetyl-2-chloropyridine ≥97.4%

Synonyms: 1-(2-chloropyridin-4-yl)ethan-1-one, 1-(2-chloropyridine-4-yl)ethanone

02228-1 02228-2 02228-3
103534-662EA 61.54 USD
103534-662 103534-664 103534-666
4-Acetyl-2-chloropyridine ≥97.4%
4-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.58 g/mol
MDL Number: MFCD07699406
CAS Number: 23794-15-2

Order Now

Specification Test Results

Purety 97.4%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR